Gene Transcripts

Gene Symbol
Olr1696
Entrez Gene ID
294158
Gene Name
olfactory receptor 1696
Gene Aliases
SCR D-8
Chromosome Location
Chr. 20: 878460 - 879425 on Build Rnor_6.0
UniGene ID
Rn.107045
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206850.1 NP_996732.1 1 633
GenBank mRNA AF034896.1 - 855

View Full Product Details