Gene Transcripts

Gene Symbol
Olr41
Entrez Gene ID
293195
Gene Name
olfactory receptor 41
Gene Aliases
Chromosome Location
Chr. 1: 167660313 - 167661254 on Build Rnor_6.0
UniGene ID
Rn.141933
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000126.1 NP_001000126.1 1 466

View Full Product Details