Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014877.3 | NP_055692.2 | 6 | 465 | |
XM_005257888.4 | XP_005257945.1 | 6 | 513 | ||
XM_005257889.4 | XP_005257946.1 | 6 | 513 | ||
XM_006722214.3 | XP_006722277.1 | 6 | 513 | ||
XM_006722215.3 | XP_006722278.1 | 6 | 513 | ||
XM_011525544.2 | XP_011523846.1 | 6 | 584 | ||
XM_017025477.1 | XP_016880966.1 | 6 | 513 | ||
XM_017025478.1 | XP_016880967.1 | 6 | 513 | ||
GenBank mRNA | AK310217.1 | - | 5 | 434 | |
BC056895.1 | - | 5 | 408 | ||
BC094881.1 | - | 4 | 332 | 3076113 | |
BC130582.1 | - | 6 | 445 | 40146613 | |
BC144083.1 | - | 6 | 445 | 9052603 | |
BP353830.1 | - | 6 | 465 | ||
D29677.2 | - | 5 | 423 |