Gene Transcripts

Gene Symbol
Olr1622
Entrez Gene ID
405127
Gene Name
olfactory receptor 1622
Gene Aliases
Chromosome Location
Chr. 15: 27438853 - 27439791 on Build Rnor_6.0
UniGene ID
Rn.142447
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000838.1 NP_001000838.1 1 585

View Full Product Details