Gene Transcripts

Gene Symbol
LEMD1
Entrez Gene ID
93273
Gene Name
LEM domain containing 1
Gene Aliases
CT50, LEMP-1
Chromosome Location
Chr. 1: 205381378 - 205449956 on Build GRCh38
UniGene ID
Hs.655520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001552.4 NP_001001552.3 4 372
NM_001199050.1 NP_001185979.1 6 514
NM_001199051.1 NP_001185980.1 5 391
NM_001199052.1 NP_001185981.1 3 249
NR_037583.1 4 326
XM_011510160.2 XP_011508462.1 7 602
XM_011510161.2 XP_011508463.1 7 590
XM_011510162.2 XP_011508464.1 7 582
XM_011510163.2 XP_011508465.1 6 482
XM_011510164.2 XP_011508466.1 8 669
XM_011510165.2 XP_011508467.1 8 641
GenBank mRNA AB084764.1 - 3 216
AB084765.1 - 4 293
AB085610.1 - 4 339
AB096677.1 - 6 481
AB096678.1 - 4 293
AB096679.1 - 5 358
AB096680.1 - 5 416
AB096681.1 - 4 339
AB096682.1 - 3 216
AK026628.1 - 4 339
AK302392.1 - 6 516
BC036636.2 - 6 484 5296437
DC399934.1 - 6 516

View Full Product Details