Gene Transcripts

Gene Symbol
DUSP12
Entrez Gene ID
11266
Gene Name
dual specificity phosphatase 12
Gene Aliases
DUSP1, YVH1
Chromosome Location
Chr. 1: 161749692 - 161757164 on Build GRCh38
UniGene ID
Hs.416216
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007240.2 NP_009171.1 4 676
XM_005244862.2 XP_005244919.1 4 765
GenBank mRNA AF119226.1 - 4 653
AK000259.1 - 2 90
BC006286.1 - 4 674 3958403
BT006633.1 - 4 642

View Full Product Details