Gene Transcripts

Gene Symbol
Tomm22
Entrez Gene ID
300075
Gene Name
translocase of outer mitochondrial membrane 22 homolog (yeast)
Gene Aliases
TOM22, rTOM22
Chromosome Location
Chr. 7: 120898536 - 120903684 on Build Rnor_6.0
UniGene ID
Rn.3119
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212514.2 NP_997679.1 3, 4 371
XM_006241974.1 XP_006242036.1 4, 5 460
GenBank mRNA AB162854.1 - 3, 4 358
BC098639.1 - 3, 4 371 7324771
FQ213035.1 - 1, 2 39
FQ215387.1 - 1, 2 39
FQ225246.1 - 1, 2 39
FQ227798.1 - 1, 2 39
FQ228672.1 - 1, 2 39
FQ230071.1 - 1, 2 39
FQ234651.1 - 1, 2 39

View Full Product Details