Gene Transcripts

Gene Symbol
Nol4
Entrez Gene ID
319211
Gene Name
nucleolar protein 4
Gene Aliases
1700013J13Rik, 4930568N03Rik, Gm1262
Chromosome Location
Chr. 18: 22693152 - 23041675 on Build GRCm38
UniGene ID
Mm.209896
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161483.1 NP_001154955.1 10 1476
GenBank mRNA AK029800.1 - 5 788
AK031753.1 - 3 469
AK086574.1 - 6 980
BC056377.1 - 10 1469 6850243

View Full Product Details