Gene Transcripts

Gene Symbol
RGL1
Entrez Gene ID
23179
Gene Name
ral guanine nucleotide dissociation stimulator like 1
Gene Aliases
RGL
Chromosome Location
Chr. 1: 183636047 - 183928551 on Build GRCh38
UniGene ID
Hs.497148
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297671.1 NP_001284600.1 2 279
NM_001297672.1 NP_001284601.1 2 279
NM_015149.4 NP_055964.3 3 654
XM_011509339.2 XP_011507641.1 4 700
XM_011509341.1 XP_011507643.1 3 607
XM_011509342.2 XP_011507644.1 3 607
XM_017000756.1 XP_016856245.1 2 128
GenBank mRNA AB023176.1 - 2 219
AF186779.1 - 2 240
AF186780.1 - 3 616
AK304293.1 - 2 204
AK309237.1 - 2 128
AK311701.1 - 2 125
AK311716.1 - 2 279
AL080117.1 - 1 22
BC136591.1 - 3 358 9020581
BX537416.1 - 3 594

View Full Product Details