Gene Transcripts
- Gene Symbol
- MORC2
- Entrez Gene ID
- 22880
- Gene Name
- MORC family CW-type zinc finger 2
- Gene Aliases
- CMT2Z, ZCW3, ZCWCC1
- Chromosome Location
- Chr. 22: 30925130 - 30968497 on Build GRCh38
- UniGene ID
- Hs.555918
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001303256.2 |
NP_001290185.1
|
24, 25 |
3250 |
|
NM_001303257.2 |
NP_001290186.1
|
24, 25 |
3250 |
|
NM_014941.3 |
NP_055756.1
|
25, 26 |
4116 |
|
XM_011530004.1 |
XP_011528306.1
|
24, 25 |
3475 |
|
XM_017028667.1 |
XP_016884156.1
|
24, 25 |
3475 |
|
XM_017028668.1 |
XP_016884157.1
|
25, 26 |
3570 |
|
GenBank mRNA |
AB020659.1 |
- |
25, 26 |
4030 |
|
AL133637.1 |
- |
6, 7 |
721 |
|
BC019257.1 |
- |
23, 24 |
2777 |
3503651 |
BC136782.1 |
- |
25, 26 |
2975 |
9020772 |
CR456469.1 |
- |
23, 24 |
2776 |
|
View Full Product Details