Gene Transcripts

Gene Symbol
NCSTN
Entrez Gene ID
23385
Gene Name
nicastrin
Gene Aliases
ATAG1874
Chromosome Location
Chr. 1: 160343056 - 160358952 on Build GRCh38
UniGene ID
Hs.517249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290184.1 NP_001277113.1 4, 5 585
NM_001290186.1 NP_001277115.1 3, 4 446
NM_015331.2 NP_056146.1 3, 4 446
XM_005245053.4 XP_005245110.1 3, 4 663
XM_011509363.2 XP_011507665.1 3, 4 663
GenBank mRNA AF240468.1 - 3, 4 464
AK299142.1 - 3, 4 333
AK310741.1 - 4, 5 526
AK314764.1 - 3, 4 353
AY359120.1 - 3, 4 323
BC047621.1 - 4, 5 542 4827966
D87442.1 - 3, 4 320

View Full Product Details