Gene Transcripts

Gene Symbol
PAMR1
Entrez Gene ID
25891
Gene Name
peptidase domain containing associated with muscle regeneration 1
Gene Aliases
DKFZP586H2123, FP938, RAMP
Chromosome Location
Chr. 11: 35431827 - 35530300 on Build GRCh38
UniGene ID
Hs.55044
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001991.2 NP_001001991.1 2, 3 704
NM_001282675.1 NP_001269604.1 4, 5 694
NM_001282676.1 NP_001269605.1 2, 3 704
NM_015430.3 NP_056245.2 2, 3 704
GenBank mRNA AF370388.1 - 1, 2 192
AK027841.1 - 2, 3 304
AK297092.1 - 4, 5 695
AK300185.1 - 2, 3 289
AK316188.1 - 2, 3 276
AL832391.1 - 2, 3 270
AY358346.1 - 2, 3 283
BC089434.1 - 2, 3 296 30558617
BX640676.1 - 2, 3 704

View Full Product Details