Gene Transcripts

Gene Symbol
MTG2
Entrez Gene ID
26164
Gene Name
mitochondrial ribosome associated GTPase 2
Gene Aliases
GTPBP5, ObgH1, dJ1005F21.2
Chromosome Location
Chr. 20: 62183025 - 62203568 on Build GRCh38
UniGene ID
Hs.340636
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015666.3 NP_056481.1 3 340
XM_005260393.1 XP_005260450.1 3 330
XM_006723774.3 XP_006723837.1 4 424
XM_011528776.1 XP_011527078.1 3 2344
XM_017027805.1 XP_016883294.1 2 2494
XM_017027806.1 XP_016883295.1 3 329
XM_017027807.1 XP_016883296.1 2 2494
XM_017027808.1 XP_016883297.1 3 330
GenBank mRNA AK001603.1 - 2 320
AK124572.1 - 3 2631
AK299147.1 - 2 114
AK299189.1 - 3 323
AK299726.1 - 3 323
AK316027.1 - 2 120
AK316325.1 - 2 117
BC012073.1 - 2 321
BC036716.2 - 2 314 4900170
CB127579.1 - 3 334
DA970672.1 - 3 340

View Full Product Details