Gene Transcripts

Gene Symbol
B9D1
Entrez Gene ID
27077
Gene Name
B9 domain containing 1
Gene Aliases
B9, EPPB9, MKS9, MKSR1
Chromosome Location
Chr. 17: 19335372 - 19378193 on Build GRCh38
UniGene ID
Hs.309467
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243473.2 NP_001230402.1 6 948
NM_001321214.1 NP_001308143.1 7 802
NM_001321215.1 NP_001308144.1 6 1041
NM_015681.4 NP_056496.1 7 713
XM_005256607.2 XP_005256664.1 7 719
GenBank mRNA AB030506.1 - 7 702
AK293815.1 - 6 1014

View Full Product Details