Gene Transcripts

Gene Symbol
FZD5
Entrez Gene ID
7855
Gene Name
frizzled class receptor 5
Gene Aliases
C2orf31, HFZ5
Chromosome Location
Chr. 2: 207762586 - 207769419 on Build GRCh38
UniGene ID
Hs.17631
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003468.3 NP_003459.2 2 5053
GenBank mRNA AK000125.1 - 1 256
AK024850.1 - 1 964
AL136852.1 - 1 596
BC095474.1 - 1 858
CR533442.1 - 1 229

View Full Product Details