Gene Transcripts

Gene Symbol
ANAPC7
Entrez Gene ID
51434
Gene Name
anaphase promoting complex subunit 7
Gene Aliases
APC7
Chromosome Location
Chr. 12: 110372900 - 110403730 on Build GRCh38
UniGene ID
Hs.719935
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137664.1 NP_001131136.1 2 282
NM_016238.2 NP_057322.2 2 282
XM_005253903.3 XP_005253960.1 2 260
XM_011538454.2 XP_011536756.1 2 510
XM_017019425.1 XP_016874914.1 2 282
XM_017019426.1 XP_016874915.1 2 277
XM_017019427.1 XP_016874916.1 2 1076
XM_017019428.1 XP_016874917.1 2 277
GenBank mRNA AF191340.1 - 2 179
AK021809.1 - 2 261
BC040130.1 - 2 197
BC098264.1 - 2 178
BC098295.1 - 2 178
BC098362.1 - 2 178
BC099732.1 - 2 178
BC111798.1 - 2 178
BC141848.1 - 2 232 8860346
BC148236.2 - 2 179 40024981
DB003237.1 - 2 283

View Full Product Details