Gene Transcripts

Gene Symbol
LEMD1
Entrez Gene ID
93273
Gene Name
LEM domain containing 1
Gene Aliases
CT50, LEMP-1
Chromosome Location
Chr. 1: 205381378 - 205449956 on Build GRCh38
UniGene ID
Hs.655520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001552.4 NP_001001552.3 4 421
NM_001199050.1 NP_001185979.1 6 563
NM_001199051.1 NP_001185980.1 5 440
NM_001199052.1 NP_001185981.1 3 298
NR_037583.1 4 375
XM_011510160.2 XP_011508462.1 7 651
XM_011510161.2 XP_011508463.1 7 639
XM_011510162.2 XP_011508464.1 7 631
XM_011510163.2 XP_011508465.1 6 531
XM_011510164.2 XP_011508466.1 8 718
XM_011510165.2 XP_011508467.1 8 690
GenBank mRNA AB084764.1 - 3 265
AB084765.1 - 4 342
AB085610.1 - 4 388
AB096677.1 - 6 530
AB096678.1 - 4 342
AB096679.1 - 5 407
AB096680.1 - 5 465
AB096681.1 - 4 388
AB096682.1 - 3 265
AK026628.1 - 4 388
AK302392.1 - 6 565
BC036636.2 - 6 533 5296437

View Full Product Details