Gene Transcripts

Gene Symbol
OR5T3
Entrez Gene ID
390154
Gene Name
olfactory receptor family 5 subfamily T member 3
Gene Aliases
OR11-178, OR5T3Q
Chromosome Location
Chr. 11: 56252200 - 56253222 on Build GRCh38
UniGene ID
Hs.553746
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004747.1 NP_001004747.1 1 396

View Full Product Details