Gene Transcripts

Gene Symbol
OR52B4
Entrez Gene ID
143496
Gene Name
olfactory receptor family 52 subfamily B member 4 (gene/pseudogene)
Gene Aliases
OR11-3
Chromosome Location
Chr. 11: 4367263 - 4368386 on Build GRCh38
UniGene ID
Hs.447463
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005161.3 NP_001005161.2 1 913
GenBank mRNA BC140928.1 - 1 913

View Full Product Details