Gene Transcripts

Gene Symbol
CFAP44
Entrez Gene ID
55779
Gene Name
cilia and flagella associated protein 44
Gene Aliases
WDR52
Chromosome Location
Chr. 3: 113286930 - 113445190 on Build GRCh38
UniGene ID
Hs.584936
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164496.1 NP_001157968.1 19 2682
NM_018338.3 NP_060808.2 19 2682
XM_005247617.3 XP_005247674.1 2 959
XM_006713696.2 XP_006713759.1 20 3272
XM_011512977.2 XP_011511279.1 20 3272
XM_011512978.2 XP_011511280.1 15 2169
XM_011512979.2 XP_011511281.1 14 1913
XM_011512980.2 XP_011511282.1 20 3272
XM_011512981.1 XP_011511283.1 20 3272
XM_011512982.2 XP_011511284.1 20 3272
XM_011512983.2 XP_011511285.1 20 3272
XM_011512984.2 XP_011511286.1 20 3272
XM_017006842.1 XP_016862331.1 20 3272
XM_017006843.1 XP_016862332.1 20 3272
XM_017006844.1 XP_016862333.1 19 3116
XM_017006846.1 XP_016862335.1 14 3067
XM_017006847.1 XP_016862336.1 15 2110
XM_017006848.1 XP_016862337.1 20 3272
XM_017006849.1 XP_016862338.1 20 3272
GenBank mRNA AK056472.1 - 19 2661
AL133606.1 - 1 24
BC101582.1 - 19 2640 8069088
BC113553.1 - 19 2641 8322605

View Full Product Details