Gene Transcripts

Gene Symbol
ROBO3
Entrez Gene ID
64221
Gene Name
roundabout guidance receptor 3
Gene Aliases
HGPPS, HGPS, RBIG1, RIG1
Chromosome Location
Chr. 11: 124865409 - 124881474 on Build GRCh38
UniGene ID
Hs.435621
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022370.3 NP_071765.2 11 1933
XM_017018122.1 XP_016873611.1 10 1698
GenBank mRNA AK291045.1 - 11 1867
AY509035.1 - 11 1956

View Full Product Details