Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168724.1 | NP_001162195.1 | 3 | 496 | |
NM_022918.3 | NP_075069.3 | 3 | 496 | ||
NR_033149.1 | 3 | 496 | |||
XM_017018140.1 | XP_016873629.1 | 2 | 361 | ||
GenBank mRNA | AK025757.1 | - | 2 | 110 | |
AK225364.1 | - | 2 | 110 | ||
AK291804.1 | - | 3 | 406 | ||
AK302473.1 | - | 3 | 300 | ||
BC030952.1 | - | 3 | 355 | ||
BC051462.1 | - | 3 | 317 | 5278478 | |
BX648678.1 | - | 3 | 482 |