Gene Transcripts

Gene Symbol
GPR89A
Entrez Gene ID
653519
Gene Name
G protein-coupled receptor 89A
Gene Aliases
GPHR, GPR89, GPR89B, SH120, UNQ192
Chromosome Location
Chr. 1: 145607960 - 145670977 on Build GRCh38
UniGene ID
Hs.504320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097612.1 NP_001091081.1 14 1833
NM_001097613.2 NP_001091082.2 15 1879
NR_036541.1 14 1853
XM_006711492.3 XP_006711555.1 15 1679
XM_006711493.3 XP_006711556.1 15 1811
XM_011509908.2 XP_011508210.1 15 1849
XM_011509909.2 XP_011508211.1 14 2061
GenBank mRNA AB097024.1 - 14 1774
AB097025.1 - 14 1774
AF132947.1 - 14 1759
AK001354.1 - 7 2571
AK021758.1 - 14 1823
AK094228.1 - 5 1774
AY358966.1 - 14 1808
BC003187.1 - 14 1736 3537949
BC067816.1 - 14 1792 5295014

Gene Symbol
GPR89B
Entrez Gene ID
51463
Gene Name
G protein-coupled receptor 89B
Gene Aliases
GPHR, GPR89, GPR89C, SH120, UNQ192
Chromosome Location
Chr. 1: 147928393 - 147993523 on Build GRCh38
UniGene ID
Hs.504320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016334.3 NP_057418.1 14 1803
XM_005277401.3 XP_005277458.1 15 1652
XM_005277402.4 XP_005277459.1 14 1940
XM_005277404.3 XP_005277461.1 14 1796
XM_006711377.3 XP_006711440.1 15 1839
XM_006711378.2 XP_006711441.1 15 1842
XM_011509610.2 XP_011507912.1 15 1836
XM_011509612.2 XP_011507914.1 15 1797
XM_017001448.1 XP_016856937.1 15 2339
GenBank mRNA AF151035.1 - 6 915
AL133020.1 - 14 2304
U78723.1 - 14 1762

View Full Product Details