Gene Transcripts

Gene Symbol
ARMCX1
Entrez Gene ID
51309
Gene Name
armadillo repeat containing, X-linked 1
Gene Aliases
ALEX1, GASP7
Chromosome Location
Chr. X: 101550531 - 101554700 on Build GRCh38
UniGene ID
Hs.9728
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016608.1 NP_057692.1 4 592
GenBank mRNA AB039670.1 - 4 592
AF248963.1 - 4 525
AK074785.1 - 4 542
AK222578.1 - 4 569
BC002691.2 - 4 531 3605690

View Full Product Details