Gene Transcripts

Gene Symbol
UBE2Q1
Entrez Gene ID
55585
Gene Name
ubiquitin conjugating enzyme E2 Q1
Gene Aliases
GTAP, NICE-5, PRO3094, UBE2Q
Chromosome Location
Chr. 1: 154548575 - 154558644 on Build GRCh38
UniGene ID
Hs.607928
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017582.6 NP_060052.3 5, 6 812
GenBank mRNA AK056388.1 - 1, 2 112
AK293986.1 - 5, 6 435
AY112698.1 - 5, 6 787
AY948200.1 - 5, 6 795
BC000848.1 - 1, 2 146
BC061583.1 - 1, 2 124
BC070158.1 - 4, 5 337
BC087836.1 - 3, 4 223
BC107057.2 - 6, 7 480
BC148185.2 - 5, 6 489

View Full Product Details