Gene Transcripts
- Gene Symbol
- UBE2Q1
- Entrez Gene ID
- 55585
- Gene Name
- ubiquitin conjugating enzyme E2 Q1
- Gene Aliases
- GTAP, NICE-5, PRO3094, UBE2Q
- Chromosome Location
- Chr. 1: 154548575 - 154558644 on Build GRCh38
- UniGene ID
- Hs.607928
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017582.6 |
NP_060052.3
|
6, 7 |
910 |
|
GenBank mRNA |
AK056388.1 |
- |
2, 3 |
210 |
|
AK293986.1 |
- |
6, 7 |
533 |
|
AY112698.1 |
- |
6, 7 |
885 |
|
AY948200.1 |
- |
6, 7 |
893 |
|
BC000848.1 |
- |
2, 3 |
244 |
|
BC009286.2 |
- |
1, 2 |
48 |
|
BC061583.1 |
- |
2, 3 |
222 |
|
BC070158.1 |
- |
5, 6 |
435 |
|
BC087836.1 |
- |
4, 5 |
321 |
|
BC107057.2 |
- |
7, 8 |
578 |
|
BC148185.2 |
- |
6, 7 |
587 |
|
View Full Product Details