Gene Transcripts

Gene Symbol
DIP2B
Entrez Gene ID
57609
Gene Name
disco interacting protein 2 homolog B
Gene Aliases
Chromosome Location
Chr. 12: 50504985 - 50748667 on Build GRCh38
UniGene ID
Hs.505516
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173602.2 NP_775873.2 37 4575
XM_005269044.1 XP_005269101.1 37 4578
XM_006719520.3 XP_006719583.1 38 4551
XM_006719521.3 XP_006719584.1 37 4388
XM_011538594.2 XP_011536896.1 38 4604
XM_011538595.2 XP_011536897.1 36 4436
XM_011538596.2 XP_011536898.1 37 4391
XM_017019699.1 XP_016875188.1 38 4586
XM_017019700.1 XP_016875189.1 37 4560
XM_017019701.1 XP_016875190.1 37 4557
XM_017019702.1 XP_016875191.1 35 4407
XM_017019703.1 XP_016875192.1 36 4354
XM_017019704.1 XP_016875193.1 36 4362
XM_017019705.1 XP_016875194.1 36 4344
GenBank mRNA AB040896.2 - 28 3189
AK097369.1 - 19 2131
AK126758.1 - 26 2946
BC007671.2 - 21 2499
BC030156.2 - 21 2499
BU429622.1 - 2 222

View Full Product Details