Gene Transcripts

Gene Symbol
Zap70
Entrez Gene ID
301348
Gene Name
zeta-chain (TCR) associated protein kinase 70
Gene Aliases
Srk
Chromosome Location
Chr. 9: 43331149 - 43353097 on Build Rnor_6.0
UniGene ID
Rn.17862
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012002.1 NP_001012002.1 13 1875
XM_006244761.2 XP_006244823.1 13 3014
XM_008766998.1 XP_008765220.1 13 2497
GenBank mRNA BC089855.1 - 13 1875

View Full Product Details