Gene Transcripts

Gene Symbol
Plekha3
Entrez Gene ID
295674
Gene Name
pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 3
Gene Aliases
Chromosome Location
Chr. 3: 63536166 - 63557378 on Build Rnor_6.0
UniGene ID
Rn.204132
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013077.1 NP_001013095.1 4 593
GenBank mRNA BC083759.1 - 4 593

View Full Product Details