Gene Transcripts

Gene Symbol
BEST2
Entrez Gene ID
54831
Gene Name
bestrophin 2
Gene Aliases
VMD2L1
Chromosome Location
Chr. 19: 12751691 - 12758458 on Build GRCh38
UniGene ID
Hs.435611
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017682.2 NP_060152.2 4, 5 644
XM_005259963.3 XP_005260020.1 5, 6 844
GenBank mRNA AF440756.1 - 4, 5 644
AK000139.1 - 4, 5 579
AK225232.1 - 4, 5 579
AY515705.1 - 4, 5 644

View Full Product Details