Gene Transcripts

Gene Symbol
Mgrn1
Entrez Gene ID
302938
Gene Name
mahogunin ring finger 1, E3 ubiquitin protein ligase
Gene Aliases
RGD1311862
Chromosome Location
Chr. 10: 10832573 - 10881981 on Build Rnor_6.0
UniGene ID
Rn.15530
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013964.1 NP_001013986.1 2 265
XM_006245792.2 XP_006245854.1 2 269
XM_006245793.2 XP_006245855.1 2 266
XM_006245794.2 XP_006245856.1 2 268
XM_006245795.2 XP_006245857.1 2 265
XM_006245796.2 XP_006245858.1 2 269
XM_008767485.1 XP_008765707.1 2 265
XM_008767486.1 XP_008765708.1 2 265
GenBank mRNA BC083621.1 - 2 265
HE574489.1 - 2 129
HE574490.1 - 2 129

View Full Product Details