Gene Transcripts

Gene Symbol
Samd14
Entrez Gene ID
287637
Gene Name
sterile alpha motif domain containing 14
Gene Aliases
RGD1306138
Chromosome Location
Chr. 10: 82820457 - 82838046 on Build Rnor_6.0
UniGene ID
Rn.156072
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024966.1 NP_001020137.1 10 2633
GenBank mRNA BC091329.1 - 10 2633

View Full Product Details