Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012643.3 | NP_001012661.1 | 3 | 1522 | |
XM_017026742.1 | XP_016882231.1 | 2 | 2267 | ||
XM_017026743.1 | XP_016882232.1 | 3 | 2377 | ||
GenBank mRNA | BC044311.1 | - | 3 | 1522 |