Gene Transcripts

Gene Symbol
Dmrtc1a
Entrez Gene ID
501668
Gene Name
DMRT-like family C1a
Gene Aliases
Dmrtc1
Chromosome Location
Chr. X: 72650314 - 72654258 on Build Rnor_6.0
UniGene ID
Rn.80016
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025288.1 NP_001020459.1 1 121
GenBank mRNA BC097356.1 - 1 121

Gene Symbol
LOC100909986
Entrez Gene ID
100909986
Gene Name
doublesex- and mab-3-related transcription factor C1-like
Gene Aliases
Chromosome Location
Chr. X|NW_007906403: 4969 - 10526 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003752209.3 XP_003752257.1 2 366
XM_006257731.2 XP_006257793.1 2 366

View Full Product Details