Gene Transcripts

Gene Symbol
Olr1241
Entrez Gene ID
300566
Gene Name
olfactory receptor 1241
Gene Aliases
Chromosome Location
Chr. 8: 42322813 - 42323745 on Build Rnor_6.0
UniGene ID
Rn.143529
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000449.1 NP_001000449.1 1 46

Gene Symbol
LOC100912488
Entrez Gene ID
100912488
Gene Name
olfactory receptor 147-like
Gene Aliases
Chromosome Location
Chr. 4: 1535100 - 1536032 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762662.1 XP_008760884.1 1 46

View Full Product Details