Gene Transcripts

Gene Symbol
Dnal4
Entrez Gene ID
300078
Gene Name
dynein, axonemal, light chain 4
Gene Aliases
Dnalc4
Chromosome Location
Chr. 7: 120987438 - 121000272 on Build Rnor_6.0
UniGene ID
Rn.3257
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009666.1 NP_001009666.1 4 1230
GenBank mRNA BC087688.1 - 4 1230

Gene Symbol
RGD1559781
Entrez Gene ID
294376
Gene Name
similar to Dynein, axonemal, light chain 4
Gene Aliases
Chromosome Location
Chr. 20: 23146936 - 23148227 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006256369.2 XP_006256431.1 1 1072

View Full Product Details