Gene Transcripts

Gene Symbol
Mtg2
Entrez Gene ID
296462
Gene Name
mitochondrial ribosome-associated GTPase 2
Gene Aliases
Gtpbp5
Chromosome Location
Chr. 3: 175448313 - 175464109 on Build Rnor_6.0
UniGene ID
Rn.17330
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013924.1 NP_001013946.1 6 872
XM_006235817.2 XP_006235879.1 6 1298
XM_006235818.2 XP_006235880.1 6 918
XM_006235820.2 XP_006235882.1 6 926
XM_008762485.1 XP_008760707.1 5 2432
XM_008762486.1 XP_008760708.1 6 1302
XM_008762487.1 XP_008760709.1 7 1019
XM_008762488.1 XP_008760710.1 7 1042
XM_008762489.1 XP_008760711.1 6 941
XM_008762490.1 XP_008760712.1 7 998
XM_008762491.1 XP_008760713.1 6 922
XM_008762493.1 XP_008760715.1 6 949
XM_008762494.1 XP_008760716.1 5 2432
XM_008762495.1 XP_008760717.1 5 2432
GenBank mRNA BC083707.1 - 6 872

View Full Product Details