Gene Transcripts

Gene Symbol
TMEM41B
Entrez Gene ID
440026
Gene Name
transmembrane protein 41B
Gene Aliases
Chromosome Location
Chr. 11: 9280654 - 9314768 on Build GRCh38
UniGene ID
Hs.594563
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001165030.2 NP_001158502.1 3 665
NM_015012.3 NP_055827.1 3 665
NR_028491.2 3 665
GenBank mRNA AK303339.1 - 3 356
BC035034.1 - 3 536 5259179
BC044597.1 - 3 527 4811565
BC064366.1 - 3 422
BC091524.1 - 3 464 30512350
D26067.1 - 3 471
DA149513.1 - 3 531
DR157551.1 - 3 518

View Full Product Details