Gene Transcripts

Gene Symbol
FREM1
Entrez Gene ID
158326
Gene Name
FRAS1 related extracellular matrix 1
Gene Aliases
BNAR, C9orf143, C9orf145, C9orf154, MOTA, TILRR, TRIGNO2
Chromosome Location
Chr. 9: 14734666 - 14911653 on Build GRCh38
UniGene ID
Hs.50850
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144966.5 NP_659403.4 17 3584
XM_005251382.3 XP_005251439.1 16 3832
XM_017014316.1 XP_016869805.1 16 3859
XM_017014317.1 XP_016869806.1 17 3402
XM_017014318.1 XP_016869807.1 17 4157
XM_017014319.1 XP_016869808.1 18 4061
XM_017014320.1 XP_016869809.1 17 3988
XM_017014321.1 XP_016869810.1 16 3651
XM_017014322.1 XP_016869811.1 16 3445
XM_017014323.1 XP_016869812.1 16 3118
XM_017014324.1 XP_016869813.1 16 3859
XM_017014325.1 XP_016869814.1 16 3859
XM_017014326.1 XP_016869815.1 15 2675
XM_017014327.1 XP_016869816.1 12 2044
XM_017014328.1 XP_016869817.1 16 3859
XM_017014329.1 XP_016869818.1 16 3859
XM_017014330.1 XP_016869819.1 16 3859
GenBank mRNA AB160987.1 - 15 2768
BX537574.1 - 17 3585

View Full Product Details