Gene Transcripts
- Gene Symbol
- SSX7
- Entrez Gene ID
- 280658
- Gene Name
- SSX family member 7
- Gene Aliases
- Chromosome Location
- Chr. X: 52644061 - 52654900 on Build GRCh38
- UniGene ID
- Hs.558712
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173358.2 |
NP_775494.1
|
1 |
119 |
|
View Full Product Details