Gene Transcripts
- Gene Symbol
- WDR13
- Entrez Gene ID
- 64743
- Gene Name
- WD repeat domain 13
- Gene Aliases
- MG21
- Chromosome Location
- Chr. X: 48597492 - 48605194 on Build GRCh38
- UniGene ID
- Hs.521973
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001166426.1 |
NP_001159898.1
|
1, 2 |
1130 |
|
NM_017883.4 |
NP_060353.2
|
2, 3 |
795 |
|
NR_029427.1 |
|
2, 3 |
761 |
|
XM_006724539.1 |
XP_006724602.1
|
3, 4 |
448 |
|
XM_006724541.3 |
XP_006724604.1
|
3, 4 |
414 |
|
XM_011543947.2 |
XP_011542249.1
|
3, 4 |
408 |
|
GenBank mRNA |
AF158978.1 |
- |
1, 2 |
1157 |
|
AF329819.4 |
- |
2, 3 |
795 |
|
AK314739.1 |
- |
2, 3 |
758 |
|
BC067094.1 |
- |
2, 3 |
442 |
|
BC080579.1 |
- |
2, 3 |
524 |
6644248 |
BI554303.1 |
- |
2, 3 |
500 |
|
DQ907252.1 |
- |
3, 4 |
487 |
|
DQ907253.1 |
- |
1, 2 |
802 |
|
DQ907255.1 |
- |
1, 2 |
745 |
|
View Full Product Details