Gene Transcripts

Gene Symbol
NPLOC4
Entrez Gene ID
55666
Gene Name
NPL4 homolog, ubiquitin recognition factor
Gene Aliases
NPL4
Chromosome Location
Chr. 17: 81556885 - 81637153 on Build GRCh38
UniGene ID
Hs.464333
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017921.3 NP_060391.2 3 336
XM_011524979.1 XP_011523281.1 3 331
XM_011524980.1 XP_011523282.1 3 331
XM_011524981.1 XP_011523283.1 3 331
XM_011524982.2 XP_011523284.1 3 331
GenBank mRNA AB040932.1 - 3 243
AK000664.1 - 3 245
AK294473.1 - 3 262
BC025930.1 - 3 277 5017718
DC322260.1 - 3 295
HY116973.1 - 3 338

View Full Product Details