Gene Transcripts

Gene Symbol
ZSWIM4
Entrez Gene ID
65249
Gene Name
zinc finger SWIM-type containing 4
Gene Aliases
Chromosome Location
Chr. 19: 13795424 - 13832254 on Build GRCh38
UniGene ID
Hs.466015
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023072.2 NP_075560.2 13 2316
XM_005260023.3 XP_005260080.1 14 2478
XM_017027153.1 XP_016882642.1 14 2749
XM_017027154.1 XP_016882643.1 14 2681
XM_017027155.1 XP_016882644.1 13 2582
XM_017027156.1 XP_016882645.1 13 2253
GenBank mRNA AK022283.1 - 2 99
AK024452.1 - 13 2316
AK299057.1 - 12 1854
BG764264.1 - 5 518

View Full Product Details