Gene Transcripts

Gene Symbol
Elmo1
Entrez Gene ID
140580
Gene Name
engulfment and cell motility 1
Gene Aliases
6330578D22Rik, C230095H21Rik, CED-12
Chromosome Location
Chr. 13: 20090240 - 20608353 on Build GRCm38
UniGene ID
Mm.342392
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080288.2 NP_525027.1 22 2371
NM_198093.3 NP_932761.1 7 894
GenBank mRNA AF398883.1 - 21 2035
AK220253.1 - 22 2259
BC024727.1 - 7 791
BC031782.1 - 11 1109 3990906

View Full Product Details