Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017983.2 | NP_001017983.2 | 9 | 3572 | |
NM_001168260.1 | NP_001161732.1 | 9 | 3670 | ||
GenBank mRNA | AK089177.1 | - | 2 | 1961 | |
AK140027.1 | - | 9 | 3532 | ||
AK148301.1 | - | 1 | 1372 | ||
AK170795.1 | - | 9 | 3623 | ||
AK171308.1 | - | 2 | 1961 | ||
BC086662.1 | - | 9 | 3502 | 30549783 |