Gene Transcripts

Gene Symbol
Amelx
Entrez Gene ID
11704
Gene Name
amelogenin, X-linked
Gene Aliases
ALGN, AMGL, AMGX, Amel, Amg, LRAP, M100888, Rgsc888
Chromosome Location
Chr. X: 169175044 - 169187287 on Build GRCm38
UniGene ID
Mm.391342
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081978.2 NP_001075447.1 4, 5 210
NM_009666.4 NP_033796.1 4, 5 210
GenBank mRNA AK029358.1 - 4, 5 214
BC059090.1 - 4, 5 180 30286944
CD773419.1 - 4, 5 214
D31768.1 - 4, 5 201
M10095.1 - 1, 2 18
S74899.1 - 5, 6 252

View Full Product Details