Gene Transcripts

Gene Symbol
KIAA0930
Entrez Gene ID
23313
Gene Name
KIAA0930
Gene Aliases
C22orf9, LSC3
Chromosome Location
Chr. 22: 45192242 - 45240769 on Build GRCh38
UniGene ID
Hs.592207
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009880.1 NP_001009880.1 10 2620
NM_015264.1 NP_056079.1 10 2692
XM_011530017.2 XP_011528319.1 10 2655
XM_017028672.1 XP_016884161.1 10 2664
GenBank mRNA AB023147.1 - 9 2388

View Full Product Details