Gene Transcripts

Gene Symbol
Zfp715
Entrez Gene ID
69930
Gene Name
zinc finger protein 715
Gene Aliases
2610041B18Rik, mszf15
Chromosome Location
Chr. 7: 43296522 - 43313311 on Build GRCm38
UniGene ID
Mm.45367
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027264.3 NP_081540.3 5 940
GenBank mRNA BC040201.1 - 6 1113
BC040386.1 - 4 1083 3585171
BC057313.1 - 5 934 6827768

View Full Product Details