Gene Transcripts
- Gene Symbol
- Hist3h2ba
- Entrez Gene ID
- 78303
- Gene Name
- histone cluster 3, H2ba
- Gene Aliases
- 1500011O09Rik, AI413321
- Chromosome Location
- Chr. 11: 58948911 - 58949372 on Build GRCm38
- UniGene ID
- Mm.28022
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018765.1 |
- |
1 |
505 |
|
BC051921.1 |
- |
1 |
495 |
5684450 |
View Full Product Details