Gene Transcripts

Gene Symbol
KLHL15
Entrez Gene ID
80311
Gene Name
kelch like family member 15
Gene Aliases
HEL-S-305
Chromosome Location
Chr. X: 23983716 - 24027186 on Build GRCh38
UniGene ID
Hs.495854
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030624.2 NP_085127.2 4 1369
XM_006724518.1 XP_006724581.1 3 1581
GenBank mRNA AB051464.1 - 2 866
AK057298.1 - 4 1369
BC109058.2 - 2 1113 40033682
BC109059.2 - 2 1113 40033683
EU794630.1 - 2 1113

View Full Product Details